• USA Home
  • Search Results

  • malvidin3glucosidechloride52889722878611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Malvidin 3-glucoside chloride

1 Products
Synonym:   Cyclamin chloride, Enin chloride, Malvidin 3-β-D-glucopyranoside, Malvidin-3-O-glucoside chloride, Oenin chloride
Image of Malvidin 3-glucoside chloride
CAS Number:
Empirical Formula (Hill Notation):
 C23H25ClO12
Molecular Weight:
 528.89
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More