• USA Home
  • Search Results

  • malvidinchloride3667564384511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Malvidin chloride

3 Products
Synonym:   3,4′,5,7-Tetrahydroxy-3′,5′-dimethoxyflavylium chloride, Malvidin, Oenidin chloride, Primulidin chloride, Syringidin chloride
Image of Malvidin chloride
CAS Number:
Empirical Formula (Hill Notation):
 C17H15ClO7
Molecular Weight:
 366.75
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
Show More