• USA Home
  • Search Results

  • manganeseiiacetylacetonate253151402458911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Manganese(II) acetylacetonate

2 Products
Synonym:   2,4-Pentanedione manganese(II) derivative, Manganous acetylacetonate, Mn(acac)2
Image of Manganese(II) acetylacetonate
CAS Number:
Linear Formula:
 [CH3COCH=C(O)CH3]2Mn
Molecular Weight:
 253.15
 Beilstein/REAXYS Number:
 4157965
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More