• USA Home
  • Search Results

  • manganeseiicarbonate1149559862911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Manganese(II) carbonate

1 Products
Synonym:   Manganese carbonate, Manganese(2+) carbonate, Manganous carbonate
Image of Manganese(II) carbonate
CAS Number:
Linear Formula:
 MnCO3
Molecular Weight:
 114.95
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More