• USA Home
  • Search Results

  • manganeseiichloridetetrahydrate197911344634911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Manganese(II) chloride tetrahydrate

7 Products
Image of Manganese(II) chloride tetrahydrate
CAS Number:
Linear Formula:
 MnCl2 · 4H2O
Molecular Weight:
 197.91
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  UV-Absorption Values
  Application(s)
  Solubility
  Product Line
  Physical Form
  Special Grade
  pH Value
Show More