• USA Home
  • Search Results

  • matrine2483651902811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Matrine

3 Products
Synonym:   Matridin-15-one, Sophocarpidine
Image of Matrine
CAS Number:
Empirical Formula (Hill Notation):
 C15H24N2O
Molecular Weight:
 248.36
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More