• USA Home
  • Search Results

  • mazindol284742223271911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mazindol

1 Products
Synonym:   5-(4-Chlorophenyl)-2,5-dihydro-3H-imidazo[2,1-a]isoindol-5-ol
Image of Mazindol
CAS Number:
Empirical Formula (Hill Notation):
 C16H13ClN2O
Molecular Weight:
 284.74
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More