• USA Home
  • Search Results

  • mcymene1342253577311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

m-Cymene

2 Products
Synonym:   1-Isopropyl-3-methylbenzene, 3-Isopropyltoluene
Image of <I>m</I>-Cymene
CAS Number:
Linear Formula:
 CH3C6H4CH(CH3)2
Molecular Weight:
 134.22
 Beilstein/REAXYS Number:
 1851357
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Titertest
  Assay
Show More