• USA Home
  • Search Results

  • mdad5solution1842513676542911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-MDA-D5 solution

2 Products
Synonym:   (±)-3,4-Methylenedioxyamphetamine-d5
Image of (±)-MDA-D<SUB>5</SUB> solution
CAS Number:
Empirical Formula (Hill Notation):
 C10D5H8NO2
Molecular Weight:
 184.25
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Anal. Standard Form
  Special Grade
  Package quantity
  Manufacturer designation
  Feature
Show More