• USA Home
  • Search Results

  • mebendazole295293143139711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mebendazole

4 Products
Synonym:   5-Benzoyl-2-benzimidazolecarbamic acid methyl ester, Mebendazol, Methyl N-(5-benzoyl-1H-benzimidazol-2-yl)carbamate
Image of Mebendazole
CAS Number:
Empirical Formula (Hill Notation):
 C16H13N3O3
Molecular Weight:
 295.29
 Beilstein/REAXYS Number:
 759809
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Special Grade
  Anal. Standard Form
  Physical Form
Show More