• USA Home
  • Search Results

  • mecopropp214651648477811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mecoprop-P

1 Products
Synonym:   (R)-2-(4-Chloro-2-methylphenoxy)propionic acid
Image of Mecoprop-P
CAS Number:
Empirical Formula (Hill Notation):
 C10H11ClO3
Molecular Weight:
 214.65
 Beilstein/REAXYS Number:
 5261833
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More