• USA Home
  • Search Results

  • medroxyprogesterone17acetate386527158911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Medroxyprogesterone 17-acetate

2 Products
Synonym:   17α-Acetoxy-6α-methylprogesterone, 17α-Hydroxy-6α-methyl-4-pregnene-3,20-dione 17-acetate, 6α-Methyl-17α-acetoxyprogesterone, 6α-Methyl-17α-hydroxyprogesterone acetate
Image of Medroxyprogesterone 17-acetate
CAS Number:
Empirical Formula (Hill Notation):
 C24H34O4
Molecular Weight:
 386.52
 Beilstein/REAXYS Number:
 2066112
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More