• USA Home
  • Search Results

  • mefloquinehydrochloride414775177392311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mefloquine hydrochloride

4 Products
Synonym:   (AS)-rel-a-(2R)-2-Piperidinyl-2,8-bis(trifluoromethyl)-4-quinolinemethanol monohydrochloride
Image of Mefloquine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C17H17ClF6N2O
Molecular Weight:
 414.77
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Application(s)
  Solubility
Show More