• USA Home
  • Search Results

  • melamine1261210878111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Melamine

4 Products
Synonym:   2,4,6-Triamino-1,3,5-triazine, sym-Triaminotriazine
Image of Melamine
CAS Number:
Empirical Formula (Hill Notation):
 C3H6N6
Molecular Weight:
 126.12
 Beilstein/REAXYS Number:
 124341
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Anal. Standard Form
  Application(s)
  Assay
  Physical Form
Show More