• USA Home
  • Search Results

  • melamined6132161355089511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Melamine-d6

1 Products
Synonym:   1,3,5-Triazine-2,4,6-triamine-d6
Image of Melamine-d<SUB>6</SUB>
CAS Number:
Empirical Formula (Hill Notation):
 C3D6N6
Molecular Weight:
 132.16
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More