• USA Home
  • Search Results

  • melatonin232287331411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Melatonin

4 Products
Synonym:   N-Acetyl-5-methoxytryptamine
Image of Melatonin
CAS Number:
Empirical Formula (Hill Notation):
 C13H16N2O2
Molecular Weight:
 232.28
 Beilstein/REAXYS Number:
 205542
 EC Number:
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Special Grade
  Assay
Show More