• USA Home
  • Search Results

  • melittinfromhoneybeevenom2846462044979011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Melittin from honey bee venom

2 Products
Image of Melittin from honey bee venom
CAS Number:
Empirical Formula (Hill Notation):
 C131H229N39O31
Molecular Weight:
 2846.46
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Activity Spectrum
  Physical Form
  Solubility
  Molecular weight
Show More