• USA Home
  • Search Results

  • memantinehydrochloride215764110052111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Memantine hydrochloride

3 Products
Synonym:   3,5-Dimethyl-1-adamantanamine hydrochloride, 3,5-Dimethylamantadine hydrochloride, 3,5-Dimethyltricyclo[3.3.1.13,7]decan-1-amine hydrochloride
Image of Memantine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C12H21N · HCl
Molecular Weight:
 215.76
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More