• USA Home
  • Search Results

  • menthol156278978111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Menthol

2 Products
Synonym:   2-Isopropyl-5-methylcyclohexanol, Hexahydrothymol, Menthol, Racemic menthol
Image of (±)-Menthol
CAS Number:
Empirical Formula (Hill Notation):
 C10H20O
Molecular Weight:
 156.27
 Beilstein/REAXYS Number:
 3194263
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More