• USA Home
  • Search Results

  • mercurydibromofluoresceindisodiumsalt7506512916811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mercury dibromofluorescein disodium salt

1 Products
Synonym:   2′,7′-Dibromo-5′-(hydroxymercurio)fluorescein disodium salt, Merbromin, Mercurochrome
Image of Mercury dibromofluorescein disodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C20H8Br2HgNa2O6
Molecular Weight:
 750.65
 EC Number:
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More