• USA Home
  • Search Results

  • mercuryiiacetate31868160027711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mercury(II) acetate

3 Products
Synonym:   Mercury(II) acetate
Image of Mercury(II) acetate
CAS Number:
Linear Formula:
 Hg(CH3COO)2
Molecular Weight:
 318.68
 EC Index Number
 216-491-1
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More