• USA Home
  • Search Results

  • meropenemtrihydrate4375111947856711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Meropenem trihydrate

3 Products
Synonym:   (1R,5S,6S)-2-[(3S,5S)-5-(dimethylaminocarbonyl)pyrrolidin-3-ylthio]-6-[(R)-1-hydroxyethyl]-1-methylcarbapen-2-em-3-carboxylic acid trihydrate
Image of Meropenem trihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C17H25N3O5S · 3H2O
Molecular Weight:
 437.51
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Special Grade
  Application(s)
Show More