• USA Home
  • Search Results

  • mesalamine153148957611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mesalamine

2 Products
Synonym:   5-Amino-2-hydroxybenzoic acid, 5-AS, 5-Aminosalicylic acid, Mesalamine
Image of Mesalamine
CAS Number:
Linear Formula:
 H2NC6H3-2-(OH)CO2H
Molecular Weight:
 153.14
 Beilstein/REAXYS Number:
 2090421
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
Show More