• USA Home
  • Search Results

  • mesoerythritol1221214932611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

meso-Erythritol

2 Products
Synonym:   1,2,3,4-Butanetetrol, meso-1,2,3,4-Tetrahydroxybutane, i-Erythritol
Image of <I>meso</I>-Erythritol
CAS Number:
Linear Formula:
 HOCH2[CH(OH)]2CH2OH
Molecular Weight:
 122.12
 Beilstein/REAXYS Number:
 1719753
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More