• USA Home
  • Search Results

  • mesoridazinebenzenesulfonate544753267269811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mesoridazine benzenesulfonate

1 Products
Synonym:   10-[2-(1-Methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-10H-phenothiazine
Image of Mesoridazine benzenesulfonate
CAS Number:
Empirical Formula (Hill Notation):
 C27H32N2O4S3
Molecular Weight:
 544.75
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More