• USA Home
  • Search Results

  • mespotassiumsalt233333994625311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

MES potassium salt

1 Products
Synonym:   2-(N-Morpholino)ethanesulfonic acid potassium salt, 4-Morpholineethanesulfonic acid potassium salt
Image of MES potassium salt
CAS Number:
Empirical Formula (Hill Notation):
 C6H12KNO4S
Molecular Weight:
 233.33
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
 eCl@ss:
 32129211
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Solubility
Show More