• USA Home
  • Search Results

  • messodiumsalt217227111923811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

MES sodium salt

5 Products
Synonym:   2-(N-Morpholino)ethanesulfonic acid sodium salt, 4-Morpholineethanesulfonic acid sodium salt
Image of MES sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C6H12NNaO4S
Molecular Weight:
 217.22
 Beilstein/REAXYS Number:
 3765682
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Agency/Method
  Suitability
  Biological Source
  Special Grade
  Application(s)
  Solubility
  Product Line
  Assay
Show More