• USA Home
  • Search Results

  • metforminhydrochloride16562111570411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Metformin hydrochloride

4 Products
Synonym:   1,1-Dimethylbiguanide hydrochloride, Metformin
Image of Metformin hydrochloride
CAS Number:
Linear Formula:
 NH2C(=NH)NHC(=NH)N(CH3)2 · HCl
Molecular Weight:
 165.62
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
Show More