• USA Home
  • Search Results

  • methacrylamide85107939011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methacrylamide

2 Products
Synonym:   2-Methylacrylamide, 2-Methylpropenamide, Methacrylic acid amide
Image of Methacrylamide
CAS Number:
Linear Formula:
 H2C=C(CH3)CONH2
Molecular Weight:
 85.10
 Beilstein/REAXYS Number:
 605397
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Potency
Show More