• USA Home
  • Search Results

  • methamphetamined14solution1633236204412011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Methamphetamine-D14 solution

2 Products
Image of (±)<WBR>-<WBR>Methamphetamine-<WBR>D<SUB>14</SUB> solution
CAS Number:
Empirical Formula (Hill Notation):
 C10D14HN
Molecular Weight:
 163.32
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Anal. Standard Form
  Special Grade
  Package quantity
  Manufacturer designation
  Feature
Show More