• USA Home
  • Search Results

  • methamphetamined5solution154266012488111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Methamphetamine-D5 solution

2 Products
Image of (±)<WBR>-<WBR>Methamphetamine-<WBR>D<SUB>5</SUB> solution
CAS Number:
Empirical Formula (Hill Notation):
 C10D5H10N
Molecular Weight:
 154.26
 EC Number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Anal. Standard Form
  Special Grade
  Package quantity
  Manufacturer designation
Show More