• USA Home
  • Search Results

  • methanesulfonamide9512314409011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methanesulfonamide

1 Products
Image of Methanesulfonamide
CAS Number:
Linear Formula:
 CH3SO2NH2
Molecular Weight:
 95.12
 Beilstein/REAXYS Number:
 1740835
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More