• USA Home
  • Search Results

  • methanesulfonicacid96117575211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methanesulfonic acid

4 Products
Synonym:   MSA
Image of Methanesulfonic acid
CAS Number:
Empirical Formula (Hill Notation):
 CH4O3S
Molecular Weight:
 96.11
 EC Index Number
 607-145-00-4
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Suitability
  Special Grade
Show More