• USA Home
  • Search Results

  • methanol32046756111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methanol

47 Products
Synonym:   Methyl alcohol
Image of Methanol
CAS Number:
Linear Formula:
 CH3OH
Molecular Weight:
 32.04
 Beilstein/REAXYS Number:
 1098229
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  UV-Absorption Values
  Anal. Standard Form
  Shelf Life
  Quality clarifier
  Miscellaneous Specifications
  Composite Dimension
  Refractive Index
  Purity Grade
  Density
  Suitability
  Product Line
  Restriction
  UV-Absorption Condition
  Ice Shipping
  Absorption
  Agency/Method
  Viscosity
  Solubility
  Color
  Vapor pressure
  Transition temperature
  Potency
  Application(s)
  Special Grade
  Physical Form
  Assay
Show More

Product #
Description
34860 Methanol, for HPLC, ≥99.9%,
322415 Methanol, anhydrous, 99.8%,
646377 Methanol, HPLC Plus, ≥99.9%,
650609 Methanol, HPLC Plus, ≥99.9%, poly-coated bottles,
34885 Methanol, for HPLC, gradient grade, ≥99.9%,
439193 Methanol, for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%,
270474 Methanol, for HPLC, gradient grade, 99.93%,
82762 Methanol, analytical standard,
179337 Methanol, ACS reagent, ≥99.8%,
32213 Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC),
1.06035 Methanol, hypergrade for LC-MS LiChrosolv®,
MX0486 Methanol, OmniSolv® LC-MS,
1.06007 Methanol, gradient grade for liquid chromatography LiChrosolv® Reag. Ph Eur,
900688 Methanol, UHPLC, for mass spectrometry,
1.06009 Methanol, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
179957 Methanol, Laboratory Reagent, ≥99.6%,
PHR1372 Methanol, Pharmaceutical Secondary Standard; Certified Reference Material,
1.03726 Methanol, for UHPLC-MS LiChrosolv®,
1.06018 Methanol, for liquid chromatography LiChrosolv®,
MX0482 Methanol, for Purge & Trap OmniSolv®,
1.00837 Methanol, for gas chromatography MS SupraSolv®,
494437 Methanol, BioReagent, ≥99.93%,
676780 Methanol, ACS reagent, ≥99.8%,
154903 Methanol, ACS spectrophotometric grade, ≥99.9%,
MX0488 Methanol, gradient grade OmniSolv®,
M1775 Methanol, Absolute - Acetone free,
1.06012 Methanol, anhydrous for analysis (max. 0.003% H₂O),
320390 Methanol, ACS reagent, ≥99.8%,
MX0475 Methanol, HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs,
1.06008 Methanol, EMPROVE® ESSENTIAL Ph Eur,BP,JPE,NF,
1.06011 Methanol, for gas chromatography ECD and FID SupraSolv®,
1.06002 Methanol, for spectroscopy Uvasol®,
8.22283 Methanol, EMPLURA®,
MX0487 Methanol, anhydrous Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
MX0485 Methanol, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
RTC000076 Methanol, Pharmaceutical Secondary Standard; Certified Reference Material,
557225 Methanol, for Purge & Trap SupraSolv®,
24229 Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC),
1.07018 Methanol, for analysis EMPARTA® ACS,
717975 Methanol, NMR reference standard,
900641 Methanol, anhydrous, ZerO2®, 99.8%,
MX0490 Methanol, histology grade,
MX0480 Methanol, OmniSolv® HR-GC,
1.13351 Methanol, for preparative chromatography Prepsolv,
MX0484 Methanol, pesticide residue grade OmniSolv®,
900516 Methanol, low benzene, ACS reagent, ≥99.8%,
MX0472 Methanol, anhydrous, septum-sealed bottle DriSolv,