• USA Home
  • Search Results

  • methanold33506184929211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methanol-d3

2 Products
Synonym:   1,1,1-Trideuteromethanol, Methyl-d3 alcohol
Image of Methanol-d<SUB>3</SUB>
CAS Number:
Linear Formula:
 CD3OH
Molecular Weight:
 35.06
 Beilstein/REAXYS Number:
 1732010
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Explosion limit
  Physical Form
  Application(s)
Show More