• USA Home
  • Search Results

  • methanold4360781198311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methanol-d4

15 Products
Synonym:   Methyl-d3 alcohol d, Tetradeuteromethanol
Image of Methanol-d<SUB>4</SUB>
CAS Number:
Linear Formula:
 CD3OD
Molecular Weight:
 36.07
 Beilstein/REAXYS Number:
 1733278
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Package quantity
  Additive
  Melting Point
  Explosion limit
  Isotopic Purity
Show More

Product #
Description
151947 Methanol-d4, ≥99.8 atom % D,
444758 Methanol-d4, "100%", 99.96 atom % D,
441384 Methanol-d4, ≥99.8 atom % D,
611646 Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS,
343803 Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS,
569534 Methanol-d4, anhydrous, ≥99.8 atom % D,
530530 Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS,
570729 Methanol-d4, ≥99.8 atom % D, anhydrous,
422878 Methanol-d4, 99 atom % D,
439029 Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS,
535435 Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS,
417653 Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS,
900596 Methanol-d4, reagent grade, ≥99 atom % D, ≥99% (CP),
1.06028 Methanol-D4, Methanol-D4 deuteration degree min. 99.8 % for NMR spectroscopy MagniSolv™. CAS 811-98-3, chemical formula CD₃OD, molar mass 36.07 g/mol.,
1.06025 Methanol-D4, Methanol-D4 deuteration degree min. 99.95% for NMR spectroscopy MagniSolv™. CAS 811-98-3, chemical formula CD₃OD, molar mass 36.07 g/mol.,