• USA Home
  • Search Results

  • methanolod3305145513611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methanol-OD

4 Products
Synonym:   Methan(ol-d), Methyl alcohol-OD, mono-Deuteromethanol
Image of Methanol-OD
CAS Number:
Linear Formula:
 CH3OD
Molecular Weight:
 33.05
 Beilstein/REAXYS Number:
 1730948
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
  Physical Form
  Additive
  Melting Point
  Explosion limit
  Assay
Show More