• USA Home
  • Search Results

  • methanolsolution32046756111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methanol solution

5 Products
Image of Methanol solution
CAS Number:
Empirical Formula (Hill Notation):
 CH4O
Molecular Weight:
 32.04
 Beilstein/REAXYS Number:
 3662861
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Explosion limit
  Absorption
  Transmittance
  Composite Dimension
  UV-Absorption Values
  Density
  Concentration
  Special Grade
  Additive
  Vapor pressure
  Vapor density vs air
  Melting Point
Show More