• USA Home
  • Search Results

  • methimazole114176056011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methimazole

3 Products
Synonym:   1-Methyl-2-imidazolethiol, 2-Mercapto-1-methylimidazole
Image of Methimazole
CAS Number:
Empirical Formula (Hill Notation):
 C4H6N2S
Molecular Weight:
 114.17
 Beilstein/REAXYS Number:
 108646
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID:
 eCl@ss
 39161002
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Physical Form
  Special Grade
Show More