• USA Home
  • Search Results

  • methoxsalen2161929881711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methoxsalen

1 Products
Synonym:   8-MOP, 8-Methoxypsoralen, 9-Methoxyfuro[3,2-g][1]benzopyran-7-one, Ammoidin, Methoxsalen, Xanthotoxin
Image of Methoxsalen
CAS Number:
Empirical Formula (Hill Notation):
 C12H8O4
Molecular Weight:
 216.19
 Beilstein/REAXYS Number:
 196453
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More