• USA Home
  • Search Results

  • methoxypolyethyleneglycolsuccinatenhydroxysuccinimide123457827432511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methoxypolyethylene glycol succinate N-hydroxysuccinimide

1 Products
Synonym:   O-Methyl-O′-succinylpolyethylene glycol 5,000 N-succinimidyl ester, PEG-succinimidyl succinate, mono-Methyl polyethylene glycol succinate N-succinimidyl ester
Image of Methoxypolyethylene glycol succinate <I>N</I>-hydroxysuccinimide
CAS Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More