• USA Home
  • Search Results

  • methyl10undecenoate1983011181911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl 10-undecenoate

2 Products
Synonym:   10-Undecenoic acid methyl ester, Undecylenic acid methyl ester
Image of Methyl 10-undecenoate
CAS Number:
Linear Formula:
 CH2=CH(CH2)8CO2CH3
Molecular Weight:
 198.30
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Assay
Show More