• USA Home
  • Search Results

  • methyl1butylcarbamoyl2benzimidazolecarbamate290321780435211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl 1-(butylcarbamoyl)-2-benzimidazolecarbamate

1 Products
Synonym:   Benomyl, NSC 263489
Image of Methyl 1-<WBR>(butylcarbamoyl)<WBR>-<WBR>2-<WBR>benzimidazolecarbamate
CAS Number:
Empirical Formula (Hill Notation):
 C14H18N4O3
Molecular Weight:
 290.32
 Beilstein/REAXYS Number:
 825455
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More