• USA Home
  • Search Results

  • methyl2bromopropionate16700544517011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl 2-bromopropionate

1 Products
Image of Methyl 2-bromopropionate
CAS Number:
Linear Formula:
 CH3CHBrCO2CH3
Molecular Weight:
 167.00
 Beilstein/REAXYS Number:
 1720592
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More