• USA Home
  • Search Results

  • methyl2pyrrolylketone10913107283911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl 2-pyrrolyl ketone

1 Products
Synonym:   2-Acetylpyrrole, Methyl 2-pyrrolyl ketone
Image of Methyl 2-pyrrolyl ketone
CAS Number:
Empirical Formula (Hill Notation):
 C6H7NO
Molecular Weight:
 109.13
 Beilstein/REAXYS Number:
 1882
 Council of Europe no.:
 4035
 EC Number:
 FEMA Number:
 3202
 Flavis number:
 14.047
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More