• USA Home
  • Search Results

  • methyl35dinitrobenzoate22614270258111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl 3,5-dinitrobenzoate

2 Products
Synonym:   3,5-Dinitrobenzoic acid methyl ester, NSC 7317
Image of Methyl 3,5-dinitrobenzoate
CAS Number:
Linear Formula:
 (O2N)2C6H3CO2CH3
Molecular Weight:
 226.14
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More