• USA Home
  • Search Results

  • methyl3aminocrotonate115131420539111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl 3-aminocrotonate

2 Products
Synonym:   Methyl 3-amino-2-butenoate
Image of Methyl 3-aminocrotonate
CAS Number:
Linear Formula:
 CH3C(NH2)=CHCOOCH3
Molecular Weight:
 115.13
 Beilstein Registry Number:
 956592
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More