• USA Home
  • Search Results

  • methyl3hydroxybutyrate11813148749611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl 3-hydroxybutyrate

2 Products
Image of Methyl 3-hydroxybutyrate
CAS Number:
Linear Formula:
 CH3CH(OH)CH2COOCH3
Molecular Weight:
 118.13
 EC Number:
 FEMA Number:
 4450
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More