• USA Home
  • Search Results

  • methyl4hydroxybenzoate152159976311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl 4-hydroxybenzoate

7 Products
Synonym:   p-Hydroxybenzoic acid methyl ester, Methyl paraben, NIPAGIN
Image of Methyl 4-hydroxybenzoate
CAS Number:
Linear Formula:
 HOC6H4CO2CH3
Molecular Weight:
 152.15
 Beilstein/REAXYS Number:
 509801
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Biological Source
  Special Grade
  Solubility
  Product Line
  Agency/Method
Show More