• USA Home
  • Search Results

  • methyl4nitrobenzoate1811561950111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl 4-nitrobenzoate

1 Products
Image of Methyl 4-nitrobenzoate
CAS Number:
Linear Formula:
 O2NC6H4CO2CH3
Molecular Weight:
 181.15
 Beilstein/REAXYS Number:
 1912198
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More